Learn More
Epredia™ HistoGel™ Specimen Processing Gel
Encapsulate and retain even small, friable and viscous specimens during histological processing with HistoGel™ Specimen Processing Gel.
Brand: Epredia™ HG-4000-012
276.50 EUR valid until 2024-12-31
Use promo code "21745" to get your promotional price.
This item is not returnable.
View return policy
Description
- Penetrates specimen without allowing specimen to escape from gel
- Outperforms other agarose gel mixtures or thromboplastin at a fraction of the cost
- Eliminates unwanted discoloration around specimens on slides
- Remains inside theparaffin block during cutting and virtually unnoticeable during sectioning
Specifications
Vial | |
Specimen Processing Gel | |
12 Pk. |