Learn More
Epredia™ Mounting Medium
Store slides for prolonged periods of time without fading of stains with Epredia™ Mounting Medium.
Brand:  Epredia™ 4111
97.75 EUR valid until 2024-12-31
Use promo code "21628" to get your promotional price.
 View more versions of this product
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 12657746
  • 118.00€ / 940mL
Estimated Shipment: 19-02-2025
Description
  • Rapid drying
  • Toluene-based
  • Optically clear
Ordering Alerts: Packaged in two configurations for convenience: a 1-pt. regular bottle or a 4-oz. plastic squeeze bottle with a twist-open/twist-close cap.
Specifications
Specifications
1 pt.
2/Cs.
Mounting Medium
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Respiratory sensitiser Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.
  • H226-Flammable liquid and vapour.
  • H336-May cause drowsiness or dizziness.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : toluene, acrylic resin