missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rifampin, 100.3%, MP Biomedicals™
Shop All MP Biomedical Products
Click to view available options
Quantity:
1 g
5 g
25 g
Specifications
Specifications
Type | Rifampicin |
Physical Form | Powder |
Color | Orange/Red |
Quantity | 25 g |
Assay Percent Range | 1.003 |
Formula Weight | 823 |
Chemical Name or Material | Rifampin |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling

Product Identifier
- Rifampin
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : Rifampin
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Rifampin, 100.3%, MP Biomedicals™ >
Spot an opportunity for improvement?Share a Content Correction