Learn More
Carl Zeiss™ Ring Illuminator
Brand:  Carl Zeiss™ 435540-9006-000
Product Code. 11825450
  • 1105.00€ / Each
Estimated Shipment: 28-07-2025
Specifications
Ring Illuminator
66 mm
1 Ea.
Product Title
Carl Zeiss™ Ring Illuminator > Diameter: 66mm