missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Chemical Name or Material | 3-(Benzoylamino)-4-methyl-N-[3-(1-methyl-1H-imidazol-2-yl)phenyl]benzamide |
---|---|
CAS | 1290490-78-6 |
Target | RIP Kinase Inhibitors |
Molecular Formula | C25H22N4O2 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18749266
|
Tocris Bioscience™
6603/10 |
10 mg |
N/A
|
N/A | |||||
18707626
|
Tocris Bioscience™
6603/50 |
50 mg |
N/A
|
N/A | |||||
Description
RIPK2 inhibitor 1 is a potent receptor-interacting protein kinase 2 (RIPK2) inhibitor (IC50 of 5-10 nM). It inhibits NF-κB activity and decreases cancer cell proliferation by 70%. In vivo, RIPK2 inhibitor 1 inhibits intestinal and lung inflammation with low toxicity in rodent models.Specifications
3-(Benzoylamino)-4-methyl-N-[3-(1-methyl-1H-imidazol-2-yl)phenyl]benzamide | |
1290490-78-6 | |
RIP Kinase Inhibitors | |
C25H22N4O2 |
Safety and Handling
Recommended Storage : Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.