missing translation for 'onlineSavingsMsg'
Learn More

Robenidine hydrochloride, 98%, Thermo Scientific™

Product Code. 15512373
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 25875-50-7
Molecular Formula C15H14Cl3N5
Molecular Weight (g/mol) 370.66
MDL Number MFCD01724404
InChI Key LTWIBTYLSRDGHP-IQGUDRQUSA-N
Synonym robenidine hydrochloride, robenidine hcl, unii-8stt15y392, 1,3-bis 4-chlorobenzylidene amino guanidine monohydrochloride, c15h13cl2n5.hcl, 1,3-bis e-4-chlorophenyl methylideneamino guanidine;hydrochloride, chimcoccid hydrochloride, khimcoccid hydrochloride, 1,3-bis 4-chlorobenzylideneamino guanidine hydrochloride
PubChem CID 16212175
IUPAC Name 1,2-bis[(E)-(4-chlorophenyl)methylideneamino]guanidine;hydrochloride
SMILES [Cl-].N\C(N[NH+]=CC1=CC=C(Cl)C=C1)=N/N=C/C1=CC=C(Cl)C=C1

Specifications

Melting Point 252.0°C to 254.0°C
Quantity 1 g
Infrared Spectrum Authentic
Packaging Glass Bottle
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 370.66
Percent Purity 98%
Chemical Name or Material Robenidine hydrochloride
compliance-icons
Product Identifier
  • Robenidine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.