Learn More
Rogo Sampaic™ Stainless Steel Rochester-Pean Forceps
For clamping larger tissue and vessels for homeostasis. Rogo Sampaic™ Stainless Steel Rochester-Pean Forceps are excellent for general laboratory use.
Brand:  Rogo Sampaic™ FML008
Product Code. 11738541
  • 172.00€ / Pack of 10
Estimated Shipment: 23-04-2025
Description
  • 160mm in length
  • Supplied in packs of 10
Specifications
Specifications
Rochester-Pean Crimp
160 mm
Product Title
Rogo Sampaic™ Stainless Steel Rochester-Pean Forceps > Length: 160mm