Learn More
Horst™ Rod Sensor
Brand:  Horst™ 70201
Product Code. 10280012
  • 116.00€ / Each
Estimated Shipment: 20-05-2025
Specifications
Rod Sensor
250 mm
Product Title
Horst™ Rod Sensor > Length: 250mm