Learn More
IKA™ Rotary Evaporator Lip Seal
For use with RV 10.8001 Rotary Evaporators.
Brand: IKA™ 0003907000
Description
- High chemical resistance
- Manufactured from a PTFE compound
- With a built-in stainless steel spring
Specifications
Lip seal | |
Rotary Evaporators Lip-seal | |
Stainless Steel Spring |
PTFE | |
RV 10.8001 rotary evaporators | |
1 Pack |
Promotions
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24156Expires: 2025-06-30
Days
36Hours
13Minutes
39Seconds
44
Product Content Correction
Product Title
IKA™ Rotary Evaporator Lip Seal > For RV 10.8001