Learn More
IKA™ ROTAVISC Standard Spindle
Standard spindle designed for use with IKA™ ROTAVISC Viscometers.
Brand: IKA™ 0025006831
This item is not returnable.
View return policy
Description
- Quick coupling
- Hook hitch
- M2.5 left-handed thread
Specifications
Standard Spindle | |
IKA™ ROTAVISC Viscometer | |
SP-4 Standard Spindle | |
AISI 316L |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Product Title
IKA™ ROTAVISC Standard Spindle > Standard Viscometer Spindle; Description: SP-4 Standard Spindle