Learn More
OHAUS™ Rotor, Angle, 24 x 1.5/2.0ml, ID
Angle Rotor for use with Frontier 5000 Series Multi Pro Centrifuges
Brand: OHAUS™ 30314838
Requires : Frontier 5000 Series Multi Pro Centrifuge
Description
- Capacity: 24 x 1.5/2.0 ml
- Max g-force: 24,325g
- Max speed: 16,000 rpm
Services
2 years
Specifications
Rotor, Angle, 24x1.5/2.0ml, ID | |
24 x 1.5/2.0 mL | |
16,000 rpm | |
FC5714 | |
NA |
Stainless Steel | |
24,325 x g | |
1.640 kg | |
45° |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
OHAUS™ Rotor, Angle, 24 x 1.5/2.0ml, ID > Model number R-A24 X 2/16MI