Learn More
Emma Safety Footwear Roy All-Round Protection Versatile XD-Fit Shoes
Designed with wipe-clean uppers and easy-clean sole.
Brand:  Emma Safety Footwear MM70156038
 View more versions of this product
Product Code. 10113744
  • 104.00€ / 1 pair
Estimated Shipment: 14-02-2025
Description
  • Full grain leather uppers
  • Hydro-Tec™ sanitized silver lining
  • Non-woven antistatic insole
  • GripForce™ Easy Twist outsole
Specifications
Specifications
Hydro-Tec Sanitized Silver lining
Non-woven antistatic insole
Steel Toe
38
GripForce™ Easy Twist outsole
All-Round Protection Versatile Shoes