missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals RPMI 1640 (1X Solution) With L-Glutamine and Sodium Bicarbonate

A general purpose medium that supports a wide variety of cells with and without supplementation.

Brand:  MP Biomedicals 091260354X6

Product Code. 15769170

This item is not returnable. View return policy

Description

Description

  • Originally developed by Moore and his colleagues at Roswell Park Memorial Institute (RPMI).
  • Based on the RPMI 1630 line of media which utilized a bicarbonate buffering system and alterations in the amounts of amino acids and vitamins.
  • RPMI has successfully been used for the cultivation of normal human and neoplastic leukocytes.
  • Popular general purpose medium when properly supplemented.
  • As a modification of McCoy′s 5A Medium, it was formulated to support lymphoblastoid cells in suspension culture, but it has since been shown to support a wide variety of cells that are anchorage dependent.
  • Originally intended to be used with a serum supplement, RPMI 1640 has been shown to support several cell lines in the absence of serum.
  • Widely used in fusion protocols and in the growth of hybrid cells.
  • Key applications: mammalian cell culture.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Clear, dark peach
Mammalian Cells
Solution
Mammalian cell culture
6 x 500 mL
Store at 2-8°C
1 X
Sterile
6.9 to 7.4
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MP Biomedicals RPMI 1640 (1X Solution) With L-Glutamine and Sodium Bicarbonate > 6 x 500 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.