missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ RSV Fusion Protein Antibody Pair [HRP]

Rabbit Monoclonal Antibody Pair

450.00€ - 1090.00€

Specifications

Assay Range 125 pg/mL
Assay Sensitivity 125-8000 pg/mL
Assay Sandwich ELISA
Conjugate HRP
Product Type Antibody Pairs
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18228097
View Documents
Novus Biologicals™
NBP2-79335-15PLATES
15 Plates
1090.00€
Pack of 15
Please to purchase this item. Need a web account? Register with us today!
18603051
View Documents
Novus Biologicals™
NBP2-79335-5Plates
5 Plates
450.00€
Pack of 5
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human respiratory syncytial virus (RSV) Fusion Protein.
Specifications

Specifications

125 pg/mL
Sandwich ELISA
Antibody Pairs
Virus
Rabbit
RUO
RSV Fusion Protein
125-8000 pg/mL
HRP
This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Human respiratory syncytial virus (RSV) Fusion Protein.
Sandwich ELISA
  • Detection Antibody: 0.2 mg/mL rabbit anti-RSV Fusion Protein monoclonal antibody conjugated to horseradish-peroxidase (HRP) (in PBS, 50 % HRP-Protector, pH 7.4).
  • Capture Antibody: 1.0 mg/mL of rabbit anti-RSV Fusion Protein monoclonal antibody (in PBS, pH 7.4).
  • Standard: Each vial contains 230 ng of recombinant RSV Fusion Protein.
  • Storage is content dependent.
    Videos
    SDS
    Documents

    Documents

    Product Certifications
    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    Novus Biologicals™ RSV Fusion Protein Antibody Pair [HRP]

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.