Learn More
Duran™ Rubber Blowers
Brand: Duran™ 292450108
30.45 EUR valid until 2025-07-13
Use promo code "24135" to get your promotional price.
This item is not returnable.
View return policy
Description
Specifications
Rubber Blower | |
For automatic burette |
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
1Hours
0Minutes
34Seconds
46 -
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
Days
73Hours
23Minutes
25Seconds
13New Offer Save Up to 85%
Product Content Correction
Product Title
Duran™ Rubber Blowers > For automatic burette