Specifications
Lens Coating | Anti-fog, Scratch-resistant |
Lens Tint | Smoke |
Product Type | Rush RUSHPSF Safety Spectacle |
No. per Pack | 1 Ea. |
Weight (Metric) | 25 g |
Documents
Product Information
Specification Sheets
RUSH
File size: 844.9 kB
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
Product Content Correction
Product Title
Bollé Safety Rush Smoke Safety Glasses >