Learn More
(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid, 97% Product Code.: 11333526
(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid, 97%
Product Code. 11333526
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
Product Code. 11333526
Brand: Thermo Scientific Alfa Aesar L14422.06
Request Bulk or Custom Format
CAS: 74163-81-8 | C10H11NO2 | 177.20 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 74163-81-8
Molecular Formula C10H11NO2
Molecular Weight (g/mol) 177.20
MDL Number MFCD00144533
InChI Key BWKMGYQJPOAASG-VIFPVBQESA-N
Synonym l-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, s-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, h-tic-oh, l-porretine, 3s-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, s-1,2,3,4-tetrahydro-3-isoquinolinecarboxylic acid, unii-737g46u1np, s---1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid, 3s-1,2,3,4-tetrahydro-isoquinoline-3-carboxylic acidShow MoreShow Less
PubChem CID 2733226
IUPAC Name (3S)-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid
SMILES OC(=O)[C@@H]1CC2=CC=CC=C2CN1
Specifications
Melting Point >300°C (decomposition)
Quantity 5 g
Beilstein 4842199
Optical Rotation −163° (c=2 in 1N NaOH)
Formula Weight 177.2
Percent Purity 97%
Chemical Name or Material (S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY08J010 Certificate TypeCertificate of Analysis Date13/11/2023 Catalog NumberL14422.06,L14422.03,L14422.14
Lot NumberM25I011 Certificate TypeCertificate of Analysis Date24/02/2023 Catalog NumberL14422.06,L14422.03,L14422.14
Lot NumberS27I035 Certificate TypeCertificate of Analysis Date04/07/2022 Catalog NumberL14422.06,L14422.03,L14422.14
Lot Number90008529 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL14422.06,L14422.03,L14422.14
Lot NumberLF004051 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL14422.06,L14422.03,L14422.14
compliance-icons
Product Identifier
  • (S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : (S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid
Product Title
(S)-(-)-1,2,3,4-Tetrahydroisoquinoline-3-carboxylic acid, 97% >