missing translation for 'onlineSavingsMsg'
Learn More
(S)-(-)-2-Amino-3-benzyloxy-1-propanol, 98+% Product Code.: 15458937

(S)-(-)-2-Amino-3-benzyloxy-1-propanol, 98+%

Product Code. 15458937
1 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15458937

Brand: Thermo Scientific Alfa Aesar H52304.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 58577-88-1 | C10H15NO2 | 181.235 g/mol

(S)-(-)-2-Amino-3-benzyloxy-1-propanol is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(-)-2-Amino-3-benzyloxy-1-propanol is used as an organic chemical synthesis intermediate.

Notes
Air sensitive. Incompatible with oxidizing agents and carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58577-88-1
Molecular Formula C10H15NO2
Molecular Weight (g/mol) 181.235
MDL Number MFCD04112477
InChI Key ZJUOMDNENVWMPL-JTQLQIEISA-N
Synonym s-2-amino-3-benzyloxy-1-propanol, h-d-ser bzl-ol, 2s-2-amino-3-benzyloxy propan-1-ol, s-2-amino-3-benzyloxy propan-1-ol, 1-propanol, 2-amino-3-phenylmethoxy-, 2s, s---2-amino-3-benzyloxy-1-propanol, 98+%, ambotzhal1035, s---2-amino-3-benzyloxy-1-propal, s---2-amino-3-benzyloxy-1-propanol
PubChem CID 11401131
IUPAC Name (2S)-2-amino-3-phenylmethoxypropan-1-ol
SMILES C1=CC=C(C=C1)COCC(CO)N

Specifications

Quantity 1 g
UN Number UN3267
Sensitivity Air Sensitive
Formula Weight 181.23
Percent Purity ≥98%
Chemical Name or Material (S)-(-)-2-Amino-3-benzyloxy-1-propanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(S)-(-)-2-Amino-3-benzyloxy-1-propanol, 98+% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.