Learn More
(S)-AMPA, Tocris Bioscience™
Product Code. 30002210
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Selective AMPA agonist; active isomer of (RS)-AMPA (Cat. No. 0169)
Active enantiomer of AMPA (EC50 = 3.5 μM).
NPEC-caged-(S)-AMPA, Inactive Enantiomer and Racemate also available.
Chemical Identifiers
CAS 83643-88-3
Molecular Formula C7H10N2O4
Molecular Weight (g/mol) 186.167
InChI Key UUDAMDVQRQNNHZ-YFKPBYRVSA-N
Synonym s-ampa, s-alpha-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid, ampa, l, unii-9280sc28gd, s-2-amino-3-3-hydroxy-5-methyl-isoxazol-4-yl-propionic acid, amq, glutamate-ampa, 1ftm, tocris-0169, tocris-0254
PubChem CID 158397
IUPAC Name (2S)-2-amino-3-(5-methyl-3-oxo-1,2-oxazol-4-yl)propanoic acid
SMILES CC1=C(C(=O)NO1)CC(C(=O)O)N
Specifications
Quantity 10 mg
Formula Weight 186.17
Percent Purity >99%
Chemical Name or Material (S)-AMPA