missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S protein RBD anti-SARS-CoV-2, Clone: AM180, ACROBiosystems
HEK293 Monoclonal Antibody
496.44€ - 3631.86€
Specifications
Antigen | S protein RBD |
---|---|
Clone | AM180 |
Applications | ELISA |
Conjugate | Unconjugated |
Form | Powder |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17206550
|
ACROBiosystems
SPD-M180-100ug |
100 μg |
496.44€
100µg |
Estimated Shipment: 09-01-2025
Log in to see stock. |
|||||
17216550
|
ACROBiosystems
SPD-M180-1mg |
1 mg |
3631.86€
1mg |
Estimated Shipment: 09-01-2025
Log in to see stock. |
|||||
Description
This product is a specific antibody against SARS-CoV-2 Spike protein RBD domain. Cross-reactivity with RBD of other coronaviruses has not been tested.Specifications
S protein RBD | |
ELISA | |
Powder | |
RUO | |
Lyophilized from 0.22μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization. | |
Primary | |
This product is a specific antibody against SARS-CoV-2 Spike protein RBD domain. Cross-reactivity with RBD of other coronaviruses has not been tested. |
AM180 | |
Unconjugated | |
Human (HEK293) | |
SARS-CoV-2 | |
IgG1 κ | |
-20°C | |
Recombinant Antibodies |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title