Learn More
Tocris Bioscience™ S-Trityl-L-cysteine
Potent, selective inhibitor of mitotic kinesin Eg5
Brand:  Tocris Bioscience™ 2191/50
128.38 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18075612
  • 159.00€ / 50mg
Estimated Shipment: 19-12-2024
Description
S-Trityl-L-cysteine is a potent, cell-permeable, selective inhibitor of mitotic kinesin Eg5, a protein required for establishing and maintaining a bipolar spindle. Inhibits basal ATPase activity (IC50 = 1 mM) and microtubule-activated ATPase activity of Eg5 (IC50 = 140 nM). Induces mitotic arrest in HeLa cells with an IC50 of 700 nM. Displays antitumor activity.
Specifications
Specifications
50 mg
S-Trityl-L-cysteine
363.47
S-Trityl-L-cysteine