missing translation for 'onlineSavingsMsg'
Learn More

S100 calcium binding protein A14 Antibody (S100A14/9077R), Alexa Fluor™ 647, Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-24305AF647

Product Code. 18692090

  • 682.00€ / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

S100 calcium binding protein A14 Monoclonal antibody specifically detects S100 calcium binding protein A14 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

S100 calcium binding protein A14
Monoclonal
Alexa Fluor 647
protein S100-A14, S100 calcium binding protein A14, S100 calcium-binding protein A14, S100A15BCMP84breast cancer membrane protein 84, S114
Recombinant full-length human S100 calcium binding protein A14 protein
0.1 mL
Cancer
57402
Store at 4°C in the dark.
IgG Îş
ELISA, Immunohistochemistry (Paraffin)
S100A14/9077R
50mM Sodium Borate
Rabbit
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
S100 calcium binding protein A14 Antibody (S100A14/9077R), Alexa Fluor™ 647, Novus Biologicals™ > 0.1 mL, Alexa Fluor 647

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.