missing translation for 'onlineSavingsMsg'
Learn More
Learn More
S100A10 Rabbit anti-Human, Clone: 6X8F7, Novus Biologicals™
Rabbit Monoclonal Antibody
196.00€ - 494.00€
Specifications
Antigen | S100A10 |
---|---|
Clone | 6X8F7 |
Dilution | Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18387023
|
Bio-Techne
NBP3-15467-20UL |
20 μg |
196.00€
20µL |
Estimated Shipment: 30-01-2025
Log in to see stock. |
|||||
18392532
|
Bio-Techne
NBP3-15467-100UL |
100 μg |
494.00€
100µL |
Estimated Shipment: 30-01-2025
Log in to see stock. |
|||||
Description
S100A10 Monoclonal antibody specifically detects S100A10 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
S100A10 | |
Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin | |
Monoclonal | |
Purified | |
RUO | |
Human | |
annexin II ligand, calpactin I, light polypeptide, ANX2L, ANX2LG, CAL1LGP11,42C, Calpactin I light chain, Calpactin-1 light chain, Cellular ligand of annexin II, CLP11Ca[1], MGC111133, p10, p10 protein, p11, protein S100-A10, S100 calcium binding protein A10, S100 calcium binding protein A10 (annexin II ligand, calpactin I, lightpolypeptide (p11)), S100 calcium-binding protein A10, S100 calcium-binding protein A10 (annexin II ligand, calpactin I, lightpolypeptide (p11)) | |
A synthetic peptide corresponding to a sequence within amino acids 1-97 of human S100A10 (P60903). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
6X8F7 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Cell Biology, Cytoskeleton Markers, Neuroscience, Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
6281 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title