Learn More
Fisherbrand™ Saddle Stool
The saddle stool enables ergonomic sitting even with frequent changes of position. Thanks to the intuitive seat height adjustment, it can be easily adapted to elevated workstations and different body heights.
Brand: Fisherbrand™ 9473L-MG02
Description
- Laboratory saddle stool
- Seat height 540-800 mm.
- Comfortable saddle seat, supports the natural S-shape of the spine.
- 3-zone upholstery foam for optimum pressure distribution.
- 3D seat dynamics to ensure active, dynamic sitting.
- Particularly small aluminium base with a diameter of 540 mm and large castors for high mobility.
- Hygienic Design.
- The stool is designed to collect no dirt and to be easy to clean.
Specifications
Saddle Stool | |
Blue | |
Steel | |
Stool |
Product Content Correction
Product Title
Fisherbrand™ Saddle Stool > Blue
Meets biosafety level S1 and S2, GMP compliant