missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Medicom™ Safetouch™ Connect Soft Latex Gloves
Click to view available options
Size:
X-Small
Medium
Large
X-Large
Description
- High elasticity and dexterity
- Watertight and airtight
- Category III
- Ambidextrous
- Recommended for medical and industrial applications
Specifications
Specifications
Certifications/Compliance | ISO 16604, ISO 10993, ISO 13485, ISO 9001 |
Certifications/Compliance (Europe) | EU Regulation 2017/745, EN 455-2, EN 455-4, EN 455-1, EN 455-3, EU regulation 2016/425, EN ISO 374-1 Type B, EN ISO 374-5:2016, EN ISO 21420 |
Accelerator Free | No |
Applications | Healthcare, Industrial |
Color | Natural |
Disposable | Single-use |
Exterior Finish | Fingertip Textured |
Finger Thickness (English) | 3.15 mil |
Finger Thickness (Metric) | 0.08 mm |
Latex Free | No |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction