Description
- Adjustable
Specifications
Specifications
Lens Coating | Anti-fog, Scratch-resistant |
Lens Tint | Clear |
Product Type | Faceshield |
Frame Color | Gray, Yellow |
Model | Tilting |
No. per Pack | 1 Ea. |
Material | Polycarbonate |
Frame Material | Polypropylene |
Documents
Product Information
Specification Sheets
PROTECTEUR RELEVABLE
File size: 600.6 kB
Special Offers
-
Multiple Manufacturers
Gear Up for Safety
Save up to 60% on gloves, apparel, and morePromo Code: 24131Expires: 2025-06-08
New Offer Shop Now
Product Content Correction
Product Title
Bollé Safety Faceshield >