Learn More
KARTELL Safety Shield
Safety Shield is free standing, portable and stables, with non-skid feet to provide facial protection from hazardous materials.
Brand:  KARTELL 0057100
94.03 EUR valid until 2025-06-08
Use promo code "24131" to get your promotional price.
 View more versions of this product
Product Code. 10061081
  • 142.00€ / Each
Estimated Shipment: 18-06-2025
Description
  • The safety shields are available in two sizes with different thicknesses and are made of high quality, clear acrylic; are free standing, portable and stable, and are all fitted with non -skid feet.
  • Art. 571 is designed to provide laboratory and healthcare personnel with facial protection from hazardous substances, aerosol and splashing.
Specifications
Specifications
14.37 in.
0.2 in.
9.53 in.
Countertop Shield
Reusable
No Frame
365 mm
5 mm
242 mm
Acrylic
None
Special Offers
  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131
    Expires2025-06-08
Product Content Correction
Product Title
KARTELL Safety Shield > Thickness: 5 mm