Learn More
Brady™ Safety Sign Hand Wash Instructions
An efficient solution with clear safety instructions to easily communicate COVID-19 countermeasures on work floors.
Brand: Brady™ 306826
17.96 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.
This item is not returnable.
View return policy
Description
- Provides clear, visual communication where it matters
- Highlights the use of appropriate personal protection
- Easily communicates social distancing and hygiene
Specifications
Laminated Polyester | |
English | |
White on Green |
Self-adhesive | |
262 x 371 mm (W x H) |