missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SAHARA S45SS Stainless Steel Heated Bath

Durable, seamless stainless steel baths for high temperature applications

Brand:  Thermo Scientific™ 1701450

Product Code. 11610941

  • 2251.20€ / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Bath volume: 30-41 L
  • Material: Stainless steel
  • Temperature: Ambient +13 to 200°C
TRUSTED_SUSTAINABILITY
Specifications

Specifications

41 L
Stainless Steel
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers