Learn More
Brannan™ Lab Thermometer with 3-points Traceable Calibration
General purpose laboratory thermometer complete with calibration certificate.
136.00€ - 246.00€
Specifications
Certifications/Compliance | UK Traceable to National Standards ISO 9001 |
---|---|
Material | Glass |
Width (Metric) | 1.2 cm |
Product Code | Brand | Temperature Range (Metric) | Thermometer Fill | Immersion Type | Length (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Temperature Range (Metric) | Thermometer Fill | Immersion Type | Length (Metric) | Price | Quantity & Availability | ||
17853048
|
Brannan™
44/406/0 + 49/101/0 |
-10°C to +150°C | Red Spirit | Partial Immersion | 30.5 cm |
140.00€
Each |
Estimated Shipment: 05-11-2024
Log in to see stock. |
||
17883008
|
Brannan™
44/002/8 + 49/101/0 |
-20°C to +110°C | Spirit | Partial Immersion | 30.5 cm |
139.00€
Each |
Estimated Shipment: 05-11-2024
Log in to see stock. |
||
17882978
|
Brannan™
44/401/0 + 49/101/0 |
-10°C to +50°C | Red Spirit | Partial Immersion | 30.5 cm |
140.00€
Each |
Estimated Shipment: 05-11-2024
Log in to see stock. |
||
17853038
|
Brannan™
44/430/0 + 49/101/0 |
-10°C to +110°C | Red Spirit | Total Immersion | 15 cm |
136.00€
Each |
Estimated Shipment: 05-11-2024
Log in to see stock. |
||
17863038
|
Brannan™
44/401/1 + 49/101/0 |
-10°C to +50°C | Red Spirit | Total Immersion | 30.5 cm |
137.00€
Each |
Estimated Shipment: 05-11-2024
Log in to see stock. |
||
17803108
|
Brannan™
44/450/0 + 49/101/0 |
-10°C to +50°C | Red Spirit | Total Immersion | 15 cm |
138.00€
Each |
Estimated Shipment: 05-11-2024
Log in to see stock. |
||
17873058
|
Brannan™
44/403/3 + 49/101/0 |
-10°C to +110°C | Green Spirit | Partial Immersion | 30.5 cm |
137.00€
Each |
Estimated Shipment: 05-11-2024
Log in to see stock. |
||
17873098
|
Brannan™
44/423/5 + 49/101/0 |
-10°C to +110°C | Green Spirit | Partial Immersion | 20.5 cm |
136.00€
Each |
Estimated Shipment: 05-11-2024
Log in to see stock. |
||
17883058
|
Brannan™
44/403/0 + 49/101/0 |
-10°C to +110°C | Red Spirit | Partial Immersion | 30.5 cm |
136.00€
Each |
Estimated Shipment: 05-11-2024
Log in to see stock. |
||
17893078
|
Brannan™
46/456/0 + 49/101/0 |
-100°C to +50°C | Red Spirit | Total Immersion | 30.5 cm |
246.00€
Each |
Estimated Shipment: 05-11-2024
Log in to see stock. |
||
17813098
|
Brannan™
44/002/7 + 49/101/0 |
-18°C to +110°C | Spirit | Partial Immersion | 30.5 cm |
139.00€
Each |
Estimated Shipment: 05-11-2024
Log in to see stock. |
||
17873118
|
Brannan™
44/010/8 + 49/101/0 |
-20°C to +110°C | Spirit | Total Immersion | 30.5 cm |
136.00€
Each |
Estimated Shipment: 05-11-2024
Log in to see stock. |
||
17883118
|
Brannan™
44/430/5 + 49/101/0 |
-10°C to +110°C | Green Spirit | Total Immersion | 15 cm |
136.00€
Each |
Estimated Shipment: 05-11-2024
Log in to see stock. |
||
Description
- Lab thermometer
- Safe non-mercury filling
- Supplied with calibration certificate
- Non-roll end cap
- Supplied as standard in protective individual, resealable telescopic tube
- Made in UK
Specifications
UK Traceable to National Standards ISO 9001 | |
Glass | |
1.2 cm |