Learn More
Spex CertiPrep™ Sample Prep Vial holder
Thick, sturdy foam block that holds 16 x standard 50mL tubes in the GenoGrinder 2010 homogeniser from SPEX SamplePrep
Brand:  Spex CertiPrep™ 2196-16-PE
Product Code. 12794127
  • 79.75€ / Each
Description
Thick, sturdy foam block that holds 16 x standard 50mL tubes in the GenoGrinder 2010 homogeniser from SPEX SamplePrep. Rugged foam block to hold sample tubes securely in 2189 clamp
Specifications
Specifications
Vial Holder
1 Pk.
Documents
One moment while we fetch your results.
Product Title
Spex CertiPrep™ Sample Prep Vial holder > Capacity: 16 x 50mL