Learn More
Velp Scientifica™ Sample Racks for Digestion Units
Facilitates nitrogen analysis and protein determination. Velp Scientifica™ Sample Racks for Digestion Units are for use with Velp Scientifica™ Kjeldahl Digestion Units.
205.00€ - 335.00€
Specifications
Product Type Sample Rack
View More Specs
Products 7
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
15379614
View Documents
Velp Scientifica™
A00000246
DKL 20
325.00€
Each
Estimated Shipment: 25-06-2025
to see stock.
Add to basket
11795654
View Documents
Velp Scientifica™
A00000168
DK20 and DKL 20 Digesters
282.00€
Each
Estimated Shipment: 05-06-2025
to see stock.
Add to basket
15309614
View Documents
Velp Scientifica™
A00000237
DK20, DKL20
208.00€
Each
Estimated Shipment: 25-06-2025
to see stock.
Add to basket
11775654
View Documents
Velp Scientifica™
A00000172
DKL12
242.00€
Each
Estimated Shipment: 20-05-2025
to see stock.
Add to basket
13487659
View Documents
Velp Scientifica™
A00000181
DKL12, 400 ml test tubes
307.00€
Each
11715664
View Documents
Velp Scientifica™
A00000180
DK 42/26 and DKl 42/26 Digesters
335.00€
Each
Estimated Shipment: 25-06-2025
to see stock.
Add to basket
11735654
View Documents
Velp Scientifica™
A00000173
DKL8
205.00€
Each
Estimated Shipment: 25-06-2025
Description
  • Available with or without heat shields
  • Available to fit a number of tube sizes and counts
Specifications
Specifications
Sample Rack
Documents
Product Certifications
Product Content Correction
Product Title
Velp Scientifica™ Sample Racks for Digestion Units