Learn More
BUCHI Sample Temperature Sensor PT1000
Sample temperature sensors monitor individual sample temperature to determine the drying endpoint.
Brand:  BUCHI 11064031
Product Code. 30130645
  • 373.00€ / Each
Estimated Shipment: 05-12-2024
Description
  • One sensor per heated plate
Specifications
Specifications
Benchtop Freeze Dryer
0
PTFE-coated Stainless Steel
220/240 V
4.5 L
Ambient to -105°C
North America