Learn More
Thermo Scientific Chemicals Sapphire substrate, 50.8mm dia x 0.332mm thick, EPI polished two sides, C-axis, HEMCOR single crystal
Brand:  Thermo Scientific Chemicals 045020.KT
 View more versions of this product
Product Code. 15463925
  • 267.33€ / Each
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sapphire Substrate, EPI Polished Two Sides
50.8 mm
50.8mm dia. x 0.332 mm thick
EPI polished two sides
1 Ea.
0.332 mm
Documents
Product Certifications
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberJ15S031 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number045019.KU,045019.KT,045020.KT
Lot NumberR21H046 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number045020.KU,045020.KT,045021.KT
Lot NumberQ23H049 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number045019.KU,045019.KT,045020.KT
Lot NumberN13A028 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number045019.KU,045019.KT,045020.KT
Lot NumberZ04F009 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number045019.KU,045019.KT,045020.KT
Product Title
Thermo Scientific Chemicals Sapphire substrate, 50.8mm dia x 0.332mm thick, EPI polished two sides, C-axis, HEMCOR single crystal > 1each