Learn More
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
11778629
|
Thermo Scientific™
DTK120R |
Chemical Trap Kit for Radioactivity |
1135.65€
Each |
In Stock
Log in to see stock. |
||||||
11708599
|
Thermo Scientific™
DC120A |
Disposable Cartridge (trapping volatile radioactivity) |
398.00€
0.25kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11738599
|
Thermo Scientific™
DC120R |
Disposable Cartridge (acid neutralization) |
380.90€
Each |
In Stock
Log in to see stock. |
||||||
15297909
|
Thermo Scientific™
ANC100 |
Acid Neutralizing Compound (acetic acid) |
283.41€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11748569
|
Thermo Scientific™
ANS121/4 |
Ammonia Neutralizing Solution (Oligo Prep) |
471.35€
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
10399102
|
Thermo Scientific™
ANS121 |
Ammonia Neutralizing Solution (Oligo Prep) |
76.98€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
12642765
|
Thermo Scientific™
SCC5 |
CryoCool Heat Transfer Fluid, 5L |
1185.90€
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11881655
|
Thermo Scientific™
LTMF190 |
Mist filter replacement cartridge | N/A | |||||||
11728669
|
Thermo Scientific™
GCF400 |
Glass condensation flask |
1001.99€
Each |
In Stock
Log in to see stock. ![]() |
||||||
10684283
|
Thermo Scientific™
CC120/DX |
Deluxe Cart |
2090.40€
Each |
Estimated Shipment: 31-07-2025
Log in to see stock. |
||||||
Description
Improve the funcionality of Thermo Scientific™ Savant™ SpeedVac™ Concentrators with Savant™ SpeedVac™ Concentrators Accessories.
Product Title
Thermo Scientific™ Savant™ DNA SpeedVac™ Concentrators Miscellaneous Accessories