Learn More
Tocris Bioscience™ sBADA
Green fluorescent D-amino acid for labeling peptidoglycans in bacteria
Brand:  Tocris Bioscience™ 7860/1
319.65 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18776836
  • 417.00€ / 1mg
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
sBADA is a green sulfonated BODIPY-FL 3-amino-D-alanine (sBADA) or fluorescent D-amino acid (FFDA). Labels peptidoglycans in live bacteria. Sulfonated form of BADA with increased hydrophilicity and thermostability. Excitation/emission λ ∽490/510 nm.
Specifications
Specifications
(R)-2-Amino-3-(3-(5,5-difluoro-7,9-dimethyl-2-sulfo-5H-4λ4,5λ4-dipyrrolo[1,2-c:2',1'-f][1,3,2]diazaborinin-3-yl)propanamido)propanoic acid
1 mg
C17H21BF2N4O6S
2376838-11-6
Fluorescent Probes for Imaging Bacteria
0.95