Learn More
GPE Scientific™ Schlenk Flask, Cylindrical with Glass Tap and Cone
GPE Scientific™ Schlenk Flask, Cylindrical with Glass Tap and Cone are specially designed for use with Schlenk lines and vacuum manifolds and are available in different capacities.
Brand:  GPE Scientific™ SCG/500
 View more versions of this product
Product Code. 15864884
  • 119.00€ / Each
Only null left
Estimated Shipment: 26-08-2025
Description
  • Cylindrical, Schlenk-style flask
  • Includes glass tap and cone
  • Available in different capacities
  • Available with different joint sizes
Specifications
Specifications
500 mL
Glass tap and cone
Cylindrical
Cylindrical
Schlenk lines and vacuum manifolds
Cylindrical Schlenk flask with tap and cone
Joint size: 24
Certificates
Promotions
Promotions
Product Content Correction
Product Title
GPE Scientific™ Schlenk Flask, Cylindrical with Glass Tap and Cone > Capacity: 500mL