Learn More
KARTELL Screw Cap Container
The Screw Cap Container has a wide mouth and comes with a screw cap lid, ideal for specimen collection.
Brand: KARTELL 0555000
This item is not returnable.
View return policy
Description
- Specimen container.
- Polyethylene cap which guarantees the perfect leakproof with milled rim allowing a safe and easy opening /closing.
- Without label.
- Excellent high and low temperatures resistance and chemical agents.
- Latex Free.
Specifications
Specimen Container | |
No | |
Cylindrical | |
1 oz. | |
Certified Quality |
Yes | |
Polypropylene | |
Non-sterile | |
30 mL |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Product Title
KARTELL Screw Cap Container > Capacity: 30 mL