Learn More
Eppendorf™ Sealing Mat for Deep Well Plates 96/1000 and 96/500
Provides secure and reliable closure and in incubation, processing, storage and archiving of samples.
Brand: Eppendorf™ 0030127978
176.00 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
- Offers high-performance sealing options for incubation and storage
- Autoclavable
Specifications
Autoclavable | |
50 | |
96/1000 and 96/500 Deep Well Plate |
96 | |
Sealing Mat, PCR Clean |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now