Learn More
BRAND™ Sealing ring for Dessicator
Provides seal for dessicator applications
Brand:  BRAND™ 65822
16.45 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10017780
  • 16.90€ / Each
Estimated Shipment: 04-06-2025
Description
  • CR construction
  • For use with Dessicators
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1
Nominal size 200 mm
For Desiccators
Desiccator Sealing Ring
Chloroprene Rubber
Documents
One moment while we fetch your results.
Product Certifications
Product Title
BRAND™ Sealing ring for Dessicator > Item Description: Nominal size 200 mm