missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Secretin Native Protein
Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: SDS-Page
Brand: Novus Biologicals™ NBP1-99449-1000UG
This item is not returnable.
View return policy
Description
Synthetic SecretinThe Secretin Native Protein has been validated for the following applications: SDS-Page.
Specifications
Lyoph | |
6343 | |
Synthetic | |
RUO | |
Secretin | |
Human | |
Sterile filtered and lyophilized with no additives |
SDS-PAGE | |
1000 μg | |
Store at -20 to -80°C. Avoid freeze-thaw cycles. | |
SCT | |
Unconjugated | |
Centrifuge the vial prior to opening. Reconstitute in sterile dH2O to a concentration >100 ug/mL. This solution can then be diluted into other aqueous buffers and stored at 4 C for 2-7 days and at -20 C for long-term storage. | |
>98%, by SDS-PAGE |