Learn More
3M™ SecureFit™ Anti-Scratch, Anti-Fog Safety Spectacles Product Code.: 15391994
Add to basket
3M™ SecureFit™ Anti-Scratch, Anti-Fog Safety Spectacles
Product Code. 15391994
Gray
Click to view available options
Lens Tint:
Amber
Blue Mirror
Clear
Gray
Indoor/Outdoor
Product Code. 15391994
Brand: 3M™ 7100078987
Edge
Add to basket
Add to basket
Reduces slippage from head movements. 3M™ SecureFit™ Anti-Scratch, Anti-Fog Safety Spectacles provide a secure and comfortable fit with the 3M Pressure Diffusion Temple Technology.
  • Available in a range of lens tints
  • Protection against UV radiation
  • Naturally adjusts to individuals' head sizes
  • Rimless lenses and fixed-length temple arm to enhance frame comfort
  • Soft, adjustable nose pads for even greater personalization of fit
  • Get added comfort over the ears with padded temple touchpoints
  • Temple arms in a bold colour for an infusion of style
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Anti-fog
Lens Tint Gray
Product Type Goggles
Lens Material Polycarbonate
  • 3M - SecureFit protective eyewear
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      3M SecureFit protective eyewear
    • 3M - SecureFit protective eyewear 600 series
      Protective eyewear that self-adjusts to individual wearer, helping to provide comfort all day.
      Video Player is loading.
      Current Time 0:00
      Duration 0:00
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 0:00
      1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
        3M SecureFit protective eyewear 600 series
      • Multiple Manufacturers
        Save up to 60% on gloves, apparel, and more
        Promo Code: 24131
        Expires2025-06-08
        New Offer
        Shop Now
      Product Content Correction
      Product Title
      3M™ SecureFit™ Anti-Scratch, Anti-Fog Safety Spectacles >