Learn More
Afora™ Basic Equipment Expansion Kit, Semi-micro
Includes 11 pieces of equipment to perform various general laboratory applications. Afora™ Basic Equipment Expansion Kit also includes a manual for each item.
Brand:  Afora™ 5002
Product Code. 11921919
  • 1490.00€ / Each
Estimated Shipment: 14-04-2025
Description
  • Includes 11 pieces for general laboratory and distillation applications
  • Box constructed of Moltopren interior housing to prevent breakage
Specifications
Specifications
  • Spherical flask: 10mL
  • Heart flask with 2 mouths: 50mL
  • Tube for calcium chloride
  • Plug
  • Willstatter Filter
  • Male threaded head
  • Propeller Shaft Rod
  • Cold finger for Claisen
  • Claisen support piece
  • Collector Separator
  • Vigreux Column: 25mL
  • Joint clips: 14/23
Distillation Apparatus
14/23