Learn More
Sensititre™ Gram Negative GNX2F AST Plate
Perform susceptibility testing of Gram-negative isolates on a single platform including colistin and Polymixin B with this research use only plate.
Brand:  Thermo Scientific GNX2F
Product Code. 17815425
  • 361.00€ / Pack of 10
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Perform susceptibility testing of non-fastidious Gram-negative isolates on a single platform with Thermo Scientific™ Sensititre™ Gram Negative GNX2F AST Plate, designed for research use only.
Decrease costs associated with multiple test methods with 21 antimicrobial compounds on a single plate, including colistin and polymixin B.
Test most bacteria, including challenging resistant organisms, on a single platform with Sensititre Gram Negative GNX2F AST Plate. 
  • Automated or manual read options are available for ultimate flexibility
  • Extended shelf life and room-temperature storage simplify inventory control
  • Individual packaging allows laboratory to test one plate at a time with no waste
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
H2O (Part No. T3339)
MIC
Sensititre Gram Negative Research Use Only MIC Plate for CRE Testing
AST Plate
Minimum Inhibitory Concentration (MIC), Carbapenem-resistant Enterobacterales (CRE)
18 hr
10
10/Pk.
For Enterobacteriaceae and Non-Enterobacteriaceae: 10μL, 30μL* (*Aid in detection of resistance mechanisms for gram negative organisms; see pack insert). For Proteus sp.: 1μL
Various
MHB (Part No. T3462)
Various
Non-fastidious, Gram-negative Organisms
35°C O2
1
Box
Autoread or manual
50 μL