Learn More
Thermo Scientific™ Sensititre™ YeastOne™ YO9 AST Plate
Simplify yeast testing with this Research Use Only Plate of nine antifungals, including Amphotericin B and Micafungin.
Brand:  Thermo Scientific™ YO9
Product Code. 17454203
  • 780.00€ / Pack of 100
Description
Simplify yeast susceptibility testing with the Thermo Scientific™ Sensititre™ YeastOne™ YO9 AST Plate with nine antifungals, including Amphotericin B and Micafungin. The Sensititre™ YeastOne™ YO9 AST Plate is intended for research use only.
Colorimetric Thermo Scientific™ alamarBlue™ agent provides reliable, easy and consistent endpoint determination with visual read option for efficient workflow and accurate results.
Achieve reliable, consistent results for Candida species with the Sensititre YeastOne YO9 AST Plate.
  • Room-temperature storage eliminates inventory control concerns
  • Individual packaging allows laboratory to test one plate at a time with no waste
  • Inclusive on-scale QC ranges provide immediate quality assurance of testing methodology
  • 24-hour incubation time enables timely results for clinicians
Not all products are available for sale in all territories. Please inquire.
VersaTREK™ and Sensititre™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
H2O (Part No. T3339)
Candida species
Sensititre YeastOne Research Use Only Plate, w/ Anidulafungin and Micafungin
Microbiology Media
35°C O2
1
10/Pk.
20µL
Various
YeastOne Broth (Part No. Y3462)
Various
Yeast Susceptibility
24-25 hr or 48-72 hr for Cryptococcus* (*For research use only. Not for use in diagnostic procedures.)
10
Manual
100 ÎĽL
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Thermo Scientific™ Sensititre™ YeastOne™ YO9 AST Plate >