Learn More
Cytiva Sentino™ Global Microbiology Peristaltic Pump
Sentino Pump replaces traditional vacuum filtration systems for membrane filtration with a small peristaltic-action pump and a sterile disposable fluid path.
Brand: Cytiva 13186
1838.25 EUR valid until 2025-04-13
Use promo code "24122" to get your promotional price.
This item is not returnable.
View return policy
Description
- The units compact design makes it easy to use in confined spaces, frees valuable bench top space and provides workspace flexibility
Specifications
NiMH nickel metal hydride battery | |
Main power supply or rechargeable battery | |
Thermoplastic Elastomer | |
24 VDC 1.6 Amp 100/240 VAC, 47/63 Hz |
Special Offers
-
Multiple Manufacturers
Precision at Your Fingertips
Master liquid handling with ease.Promo Code: 24122Expires: 2025-04-13
Product Content Correction
Product Title
Cytiva Sentino™ Global Microbiology Peristaltic Pump > 1/Ea.