missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Separating or resolving buffer (4X)
61.00€ - 97.38€
Specifications
Concentration | 4X |
---|---|
Form | Liquid |
Storage Requirements | Ambient temperatures |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15492989
|
Thermo Scientific Chemicals
J60181.AP |
500mL |
61.00€
500mL |
In Stock
Log in to see stock. |
|||||
15402999
|
Thermo Scientific Chemicals
J60181.K2 |
1L |
97.38€
1 L |
In Stock
Log in to see stock. |
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4X | |
Liquid | |
Ambient temperatures |
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title