accessibility menu, dialog, popup
Test
Search
Order Status
Quick Order
Support
Sign In
chevron-down
chevron-down
Learn More
Home
Products
Gases and Gas Accessories
Gas Accessories
Gas Cylinder Supports
Hamilton™ Septas
10208092
Print
Hamilton™ Septas
Outer Diameter: 5mm
Brand:
Hamilton™ 75829
View more versions of this product
Product Code.
10208092
114.00€
/ Pack of 25
Quantity
Estimated Shipment: 12-03-2025
Log in
to see stock.
Explore more special offers
Add to basket
This item is not returnable.
View return policy
Specifications
Diameter (Metric) Outer
5 mm
Product Type
Septa