missing translation for 'onlineSavingsMsg'
Learn More

Sertaconazole nitrate, Thermo Scientific™

Brand:  Thermo Scientific Acros 469250010

Additional Details : CAS : 99592-39-9 Weight : 0.00100kg

 View more versions of this product

Product Code. 17710649

  • 128.00€ / 1g
Estimated Shipment: 16-07-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

Sertaconazole nitrate
1 g
No special requirements
C20 H15 Cl3 N2 O S . H N O3
White to Beige
99592-39-9
156°C to 165°C
Life science
Powder
≥97.5%
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers