Learn More
Sertaconazole nitrate, Thermo Scientific™
Brand: Thermo Scientific Acros 469252500
Additional Details : CAS : 99592-39-9 Weight : 0.00025kg
Specifications
Sertaconazole nitrate | |
250 mg | |
No special requirements | |
C20 H15 Cl3 N2 O S . H N O3 | |
White to Beige |
99592-39-9 | |
156°C to 165°C | |
Life science | |
Powder | |
≥97.5% |